Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioic acid | 1268488-70-5 | MFCD11041130 | 97.0% | 514.56 | C22H42O13 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1268945G
Encompass_Preferred
Bis-PEG9-acid is a polyethylene glycol (PEG)-based bifunctional linker with two terminal carboxylic acid groups used for PROTAC synthesis and as a cleavable ADC linker in bioconjugation applications.
  • Peg-based bifunctional linker with two terminal carboxylic acid groups.
  • Suitable for targeted protein degradation (PROTAC) synthesis.
  • Usable as a cleavable antibody-drug conjugate (ADC) linker for bioconjugation.
  • Molecular weight 514.56, useful for hydrophilic spacer applications.
  • Typical purity reported ~97-98% and CAS number 1268488-70-5.
Catalog No. 50-002-02342