Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25-octaoxaoctacosanedioic acid | 1246189-43-4 | >97.0% | 470.51 g/mol | C20H38O12 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY126893500MG
Encompass_Preferred
Bis-PEG8-acid is a polyethylene glycol (PEG)-based cleavable linker containing terminal carboxylic acid groups. It is used as a hydrophilic spacer in the synthesis of proteolysis targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs), improving solubility and flexibility between conjugated moieties.
  • Cleavable PEG-based linker with terminal carboxylic acids.
  • Hydrophilic spacer that increases aqueous solubility.
  • Facilitates conjugation in PROTAC and ADC synthesis.
  • High purity suitable for research applications.
  • Supplied as a 500 mg quantity for laboratory use.
Catalog No. 50-002-04202