Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16,19,22,25-octaoxaoctacosanedioic acid | 1246189-43-4 | 98.0% | 470.51 | C20H38O12 | 10 G
SDP

Supplier:  Medchemexpress LLC HY12689310G

Encompass_Preferred

Bis-PEG8-acid is a bifunctional polyethylene glycol (PEG) linker with two terminal carboxylic acid groups used for the synthesis of PROTACs and antibody-drug conjugates. It is a white to light yellow solid with high purity and good solubility in DMSO.

  • Contains two terminal carboxylic acid groups.
  • Molecular formula C20H38O12; molecular weight 470.51.
  • Purity 98.0%.
  • Soluble in DMSO at 100 mg/mL (≈212.54 mM).
  • Storage: powder at -20°C for long term; in solvent store at -80°C for up to 6 months.
  • Available in multiple pack sizes including 10 g.

Catalog No. 50-002-04133


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.