Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25-octaoxaoctacosanedioic acid | 1246189-43-4 | 98.0% | 470.51 | C20H38O12 | 250 MG

Supplier: Medchemexpress LLC HY126893250MG
Bis-PEG8-acid is a polyethylene glycol (PEG) based bifunctional linker with terminal carboxylic acid groups used for PROTAC synthesis and as a cleavable linker in antibody-drug conjugates (ADCs).
- Provides a flexible PEG8 spacer for conjugation chemistry
- Contains terminal carboxylic acid groups for amide coupling
- Molecular formula C20H38O12 and molecular weight 470.51
- Purity 98.0%
- Available in multiple package sizes, including 250 MG
- Powder storage: -20°C (stable 3 years) or 4°C (stable 2 years)
- In solvent storage: -80°C (stable 6 months) or -20°C (stable 1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.