Learn More
Medchemexpress LLC 4,7,10,13,16,19,22,25-octaoxaoctacosanedioic acid | 1246189-43-4 | >98.0% | 470.51 g·mol⁻¹ | C20H38O12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126893100MG
Encompass_Preferred
Bis-PEG8-acid is a polyethylene glycol (PEG)-based bifunctional linker used for conjugation chemistry in the synthesis of PROTACs and cleavable antibody-drug conjugates. It provides a PEG8 spacer with terminal carboxylic acid groups to enable standard coupling reactions and is supplied for research use.
  • PEG8 spacer with terminal carboxylic acid groups.
  • Suitable for PROTAC linker synthesis and cleavable ADCs.
  • High purity; >98.0% reported by manufacturer.
  • Available in multiple pack sizes for research workflows.
Catalog No. 50-002-04174