Learn More
Medchemexpress LLC 4,7,10,13,16-pentaoxanonadec-18-ynoic acid | 1245823-51-1 | MFCD22201536 | ≥98.0% | 304.34 g·mol⁻¹ | C14H24O7 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1011575G
Encompass_Preferred
Propargyl-PEG5-acid is an alkyne-terminated polyethylene glycol linker with five ethylene glycol units used for assembling antibody-drug conjugates, PROTACs, and click-chemistry conjugations. It provides a hydrophilic spacer and reactive termini for modular bioconjugation.
  • Alkyne terminus enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click reactions.
  • Carboxylic acid terminus allows amide coupling to amines.
  • Five-unit PEG spacer increases hydrophilicity and solubility.
  • High purity (≥98.0% by NMR) suitable for synthesis applications.
  • Moderate molecular weight (304.34 g·mol⁻¹) for predictable linker length.
  • Colorless to light yellow liquid; store protected from light at low temperature.
Catalog No. 50-002-13161