Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13,16-pentaoxanonadec-18-ynoic acid, 2,5-dioxo-1-pyrrolidinyl ester | 1393330-40-9 | MFCD22574798 | 98.0% | 401.41 g/mol | C18H27NO9 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY130388500MG

Encompass_Preferred

Propargyl-PEG5-NHS ester is a bifunctional PEG linker reagent with an alkyne (propargyl) group for click chemistry and an N-hydroxysuccinimide (NHS) ester for amine conjugation. It is intended for laboratory bioconjugation workflows such as antibody-drug conjugate (ADC) and PROTAC synthesis.

  • Contains a propargyl (alkyne) functional group for CuAAC click reactions.
  • Has an NHS ester for efficient amine coupling to primary amines.
  • Molecular weight 401.41 g/mol and formula C18H27NO9.
  • Purity typically 98.0% as supplied.
  • Supplied as a solid/liquid reagent for research use with recommended cold, light-protected storage.

Catalog No. 50-002-03242


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.