Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10,13-tetraoxa-16-azaheneicosanoic acid, 21-[(3as,4s,6ar)-hexahydro-2-oxo-1h-thieno[3,4 | 1334172-58-5 | 96.2% | 657.65 g/mol | C27H36F5N3O8S | 100 MG
SDP

Supplier:  Medchemexpress LLC HY138488100MG

Encompass_Preferred

Biotin-PEG4-PFP ester is a cleavable four-unit polyethylene glycol (PEG4) biotinylation reagent bearing a pentafluorophenyl (PFP) ester reactive group used for labeling and as an antibody-drug conjugate (ADC) linker. It is supplied for research use only.

  • Cleavable PEG4 spacer increases hydrophilicity and reduces steric hindrance.
  • PFP ester reactive group enables efficient conjugation to primary and secondary amines.
  • High purity (96.18%) and available in multiple pack sizes, including 100 MG.
  • Storage: powder at -20°C; in solvent stable at -80°C for 6 months or -20°C for 1 month.
  • CAS number 1334172-58-5; molecular weight 657.65 g/mol; formula C27H36F5N3O8S.

Catalog No. 50-002-03707


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.