Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxa-16-azanonadecanoic acid, 19-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-17-oxo | 1263045-16-4 | 99.8% | 500 MG

Supplier: Medchemexpress LLC HY130414500MG
Mal-amido-PEG4-acid is an off-white to yellow solid with a molecular formula of C18H28N2O9 and a molecular weight of 416.42. It exhibits a high purity of 99.76% as determined by HPLC analysis, and its 1H NMR and LCMS spectra are consistent with the chemical structure.
- Off-white to yellow solid appearance
- Molecular formula C18H28N2O9
- Molecular weight 416.42
- Purity of 99.76% (HPLC)
- NMR and LCMS data consistent with structure
- Store at -20°C, protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.