Learn More
Medchemexpress LLC 4,7,10,13-Tetraoxapentadecan-1-ol, 15-azido- | 2028281-87-8 | ≥97.0% | C11H23N3O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140809100MG
Encompass_Preferred
Azido-PEG4-(CH2)3OH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker for chemical synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group for diverse reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Supports the creation of molecules that leverage the ubiquitin-proteasome system for protein degradation
Catalog No. 50-003-53547