Learn More
Medchemexpress LLC 4,7,10-trioxa-11-thiadodecanoic acid, 1,1-dimethylethyl ester, 11,11-dioxide | 1312309-62-8 | ≥97.0% | C12H24O7S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140383100MG
Encompass_Preferred
Ms-PEG2-C2-Boc is a PEG-based PROTAC linker. PROTACs are molecules that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • Can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-56926