Learn More
Medchemexpress LLC 4,7,10-Trioxa-11-thiadodecanoic acid, 1,1-dimethylethyl ester, 11,11-dioxide | 1312309-62-8 | ≥97.0% | C12H24O7S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140383
Encompass_Preferred
Ms-PEG2-C2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. These PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Acts as a PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • PROTACs utilize two ligands connected by a linker
  • Facilitates binding to an E3 ubiquitin ligase
  • Enables targeting of specific proteins
  • Leverages the intracellular ubiquitin-proteasome system
  • Promotes selective degradation of target proteins
Catalog No. 50-003-57226