Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | >=98.0% | 264.43 g/mol | C12H28O4Si | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0424491G

Encompass_Preferred

TBDMS-PEG3-OH is a tert-butyldimethylsilyl-protected PEG3 linker with a terminal hydroxyl group. It is supplied as a colorless to light yellow liquid and is used as a short, flexible spacer in medicinal chemistry, linker synthesis, and drug-delivery research.

  • tert-butyldimethylsilyl-protected terminal hydroxyl
  • three ethylene glycol units (PEG3)
  • molecular weight 264.43 g/mol
  • chemical formula C12H28O4Si
  • CAS number 201037-95-8
  • purity >=98.0% (1H NMR)
  • appearance colorless to light yellow liquid
  • storage: pure form -20°C for 3 years; 4°C for 2 years

Catalog No. 50-002-10663


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.