Learn More
Medchemexpress LLC 4,7,10-trioxa-3-siladodecan-12-ol, 2,2,3,3-tetramethyl- | 201037-95-8 | MFCD30471718 | ≥98.0% | 264.43 | C12H28O4Si | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0424495G
Encompass_Preferred
TBDMS-PEG3-OH is a tert-butyldimethylsilyl (TBDMS)-protected triethylene glycol (PEG3) derivative used as a short PEG linker and building block in synthetic chemistry and drug-delivery research. It is a colorless to light yellow liquid (formula C12H28O4Si, MW 264.43) with high purity and defined storage conditions.
  • Tert-butyldimethylsilyl-protected hydroxyl for selective deprotection.
  • Short PEG3 spacer ideal for linker synthesis and conjugation.
  • High purity: ≥98.0%.
  • Available in bench-scale pack sizes, including 5 G.
  • Stable when stored at recommended temperatures (pure: -20°C to 4°C).
Catalog No. 50-002-10770