Learn More
Medchemexpress LLC 4,7,12-Trioxa-10-azatetradecanoic acid, 13,13-dimethyl-11-oxo-, 2,5-dioxo-1-pyrrolidinyl ester | 2183440-73-3 | ≥98.0% | C16H26N2O8 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141117
Encompass_Preferred
Boc-NH-PEG2-C2-NHS ester is an alkyl/ether-based PROTAC linker designed for use in the synthesis of PROTACs. These compounds exploit the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins, offering a novel approach for targeted therapy.
  • Utilized in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Alkyl/ether-based linker
Catalog No. 50-003-52728