Learn More
Medchemexpress LLC 4,7-dihydro-6-(1-methylethyl)-7-oxo-5-phenyl-pyrazolo[1,5-a]pyrimidine-3-carbonitrile | 1628208-23-0 | 98.2% | 278.31 | C16H14N4O | 1 ML

Supplier: Medchemexpress LLC HY10042110MM 1ML
CPI-455 is a potent small-molecule inhibitor of KDM5 histone demethylases used in research to probe epigenetic regulation and H3K4 methylation dynamics. It is suitable for biochemical and cell-based assays investigating demethylase activity and drug-tolerant cell populations.
- Potent KDM5 inhibition with IC50 ≈ 10 nM.
- Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
- High purity suitable for research (≈98.2% reported).
- Molecular formula C16H14N4O; molecular weight 278.31 g·mol^-1.
- Soluble in DMSO for straightforward assay preparation.
- Stable when stored frozen; follow supplier storage recommendations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.