Learn More
Medchemexpress LLC 4,7-dihydro-6-(1-methylethyl)-7-oxo-5-phenyl-pyrazolo[1,5-a]pyrimidine-3-carbonitrile | 1628208-23-0 | 98.2% | 278.31 | C16H14N4O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10042125MG
Encompass_Preferred
CPI-455 is a potent pan-KDM5 (histone lysine demethylase 5) inhibitor used in research to increase H3K4 trimethylation and to study mechanisms of drug tolerance in cell models. Supplied as a white to off-white solid for in vitro and preclinical investigations, it should be handled and stored according to laboratory safety guidelines.
  • Inhibits KDM5 with low nanomolar potency.
  • Elevates global H3K4me3 levels in treated cells.
  • Reduces drug-tolerant persister cell populations in models.
  • Provided as a solid with defined stability and storage conditions.
  • Available in small research pack sizes suitable for assay work.
Catalog No. 50-002-13910