Learn More
Medchemexpress LLC 4-Acetamidobutanoic acid | MFCD00020552 | 98.0% | 145.16 g/mol | C6H11NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101411S10MG
Encompass_Preferred
4-Acetamidobutanoic acid (N-acetyl GABA) is an endogenous metabolite of GABA used in research applications. It exhibits reported antioxidant and antibacterial activities and is supplied as a stable powder with documented storage recommendations. Chemical identifiers include CAS 3025-96-5, molecular formula C6H11NO3, and molecular weight 145.16 g/mol. Typical purity is 98.0%.
  • High purity of 98.0% suitable for analytical work.
  • Available in small to bulk pack sizes, including 10 mg samples.
  • Stable as a powder under recommended cold storage.
  • Suitable for studies of GABA metabolism and related assays.
  • Includes handling guidance for stock solutions and sterilization.
Catalog No. 50-002-45619