Learn More
AdipoGen 4-Acetoxystyrene Stabil MEHQ
SDP
Supplier:  AdipoGen CDXA0708G025
Encompass_Preferred
Chemical. CAS 2628-16-2. Formula C10H10O2. MW 162.2. Styrene monomer used as building block for synthesis e.g. polymer. Precursor to p-hydroxystyrene that is readily derivatized. Undergoes free radical polymerization, similar to styrene. Contains 300 ppm MEHQ as a stabilizer.
Catalog No. 50-256-1727