Learn More
Cayman Chemical 4-Acetyl ChrysomycIn B 2.5mg
SDP
Supplier:  Cayman Chemical 330242.5 MG
Encompass_Preferred
A bacterial metabolite; active against strains of B. subtilis, B. cereus, Corynebacterium bovis, and E. faecalis (MICs = 2-64 UG/ml); cytotoxic against a panel of human cancer cell lines, including doxorubicin-sensitive or -resistant cells (IC50s = 1.7-14 and 260-370 ng/ml, respectively)
Catalog No. 50-294-8287