Learn More
Medchemexpress LLC 4'-Acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1,1'-biphenyl]-4-carboxamide | 162408-66-4 | MFCD00936840 | 99.9% | 485.62 g/mol | C30H35N3O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10138225MG
Encompass_Preferred
GR 103691 is a potent, selective dopamine D3 receptor antagonist (Ki = 0.4 nM) with more than 100-fold selectivity versus D4 and D1 receptors. It also shows affinity for 5-HT1A (Ki = 5.8 nM) and α-1 adrenoceptors (Ki = 12.6 nM). Supplied as a high-purity solid for research use.
  • Potent D3 receptor antagonist (Ki = 0.4 nM).
  • High selectivity for D3 over D4 and D1 receptors.
  • Additional affinity for 5-HT1A and α-1 adrenoceptors.
  • High reported purity (99.91%).
  • Solid, white to off-white appearance suitable for handling and storage.
  • Stable when stored as powder at -20°C for extended periods.
  • Available in small milligram package sizes for preclinical research.
Catalog No. 50-002-13213