Learn More
Medchemexpress LLC 4-Acetylbiphenyl | 92-91-1 | 99.3% | 196.24 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW009877100G
Encompass_Preferred
4-Acetylbiphenyl (ACBP) is a biphenyl compound that can be used as a probe molecule to study the structure and function of proteins and to study the properties of lipid membranes.
  • Can be used as a probe molecule to study the structure and function of proteins
  • Can be used to study the properties of lipid membranes
Catalog No. 50-004-52486