Learn More
Medchemexpress LLC 4-Acetylphenylboronic acid | 149104-90-5 | HY-32933 | 99.96% | 163.97 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3293310G
Encompass_Preferred
4-Acetylphenylboronic acid is a potent inhibitor of carbonic anhydrases II (CA II), with IC50s of 246 μM and 281.40 μM for bovine CA II (bCA II) and human CA II (hCA II). It can be obtained via mechanochemistry and solvent evaporation and exhibits a loss of the carbonyl group at 500-550 K. In vitro, 4-Acetylphenylboronic acid (30 μM) shows 18.1% inhibition of cells against the BJ Human fibroblast cell line, suggesting no cytotoxicity.
  • Potent inhibitor of carbonic anhydrases II (CA II)
  • Can be obtained via mechanochemistry and solvent evaporation
  • No significant cytotoxicity against BJ Human fibroblast cell line at 30 μM
Catalog No. 50-004-04722