Learn More
Medchemexpress LLC 4-amino-1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one | 69123-90-6 | MFCD00868897 | 99.3% | 371.10 g/mol | C9H11FIN3O4 | 10 MG

Supplier: Medchemexpress LLC HY5073510MG
Fiacitabine is a fluorinated, iodinated pyrimidine nucleoside analogue used as a research reagent and a selective inhibitor of herpes simplex virus (HSV) DNA replication. It is supplied in small quantities for in vitro studies and is identified by CAS 69123-90-6.
- Selective inhibitor of HSV DNA replication.
- Intended for in vitro research applications.
- Reported high purity (about 99.3%).
- Molecular weight 371.10 g/mol.
- Chemical formula C9H11FIN3O4.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.