Learn More
Medchemexpress LLC 4-Amino-2-fluorobenzoic acid | 446-31-1 | 155.13 | 10 G

Supplier: Medchemexpress LLC HYW00784710G
4-Amino-2-fluorobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.
- Biochemical reagent for life science research
- Molecular weight of 155.13
- CAS number 446-31-1
- Appears as a solid
- Color is light brown to yellow
- Purity of 99.9%
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.