Learn More
Medchemexpress LLC 4-amino-N-[(4R,5S)-1-azabicyclo[3.3.1]nonan-4-yl]-5-chloro-2-methoxybenzamide | 112727-80-7 | MFCD00869601 | 98.4% | 323.82 g/mol | C16H22ClN3O2 | 5 MG

Supplier: Medchemexpress LLC HY141475MG
Renzapride is a substituted benzamide research compound that acts mainly as a 5-HT4 receptor agonist and also exhibits 5-HT3 and 5-HT2b antagonist activity. It is provided for laboratory research use to support studies of gastrointestinal motility and serotonin receptor pharmacology. The compound is supplied as a solid with high purity suitable for in vitro and in vivo pharmacology assays.
- Acts as a 5-HT4 receptor agonist and 5-HT3/5-HT2b antagonist.
- Suitable for in vitro and in vivo gastrointestinal motility research.
- Supplied as a solid for laboratory use with high reported purity.
- Molecular formula C16H22ClN3O2; molecular weight 323.82 g/mol.
- Soluble in DMSO; handle and store according to supplier guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.