Learn More
Medchemexpress LLC 4-aminobenzamide | 2835-68-9 | MFCD00007999 | 98.7% | 136.15 g/mol | C7H8N2O | 1 ML

Supplier: Medchemexpress LLC HYW01704110MM/1ML
4-Aminobenzamide is a biochemical reagent used in life-science research as an organic compound and assay reagent. Supplied as a 10 mM solution in DMSO (1 mL), it is used in biochemical assays, enzyme inhibition studies, and small-molecule research. The material has a reported purity of 98.72% and a molecular weight of 136.15 g/mol.
- Provided as a ready-to-use 10 mM solution in DMSO.
- High purity suitable for research applications.
- Soluble in DMSO; ultrasonic assistance may improve solubility.
- Storage recommendations for solid and solution forms to preserve stability.
- White to off-white solid when supplied in bulk; provided as solution for the 10 mM/1 mL pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.