Learn More
Medchemexpress LLC 4-Aminobenzamide | 2835-68-9 | MFCD00007999 | 98.7% | 136.15 g/mol | C7H8N2O | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01704150G
Encompass_Preferred
4-Aminobenzamide (CAS 2835-68-9) is an organic biochemical reagent used in life-science research, commonly employed as a PARP inhibitor to study cell death and cytotoxicity. It is supplied in solid and solution formats for laboratory assays and is characterized by a defined purity and stability when stored under recommended conditions.
  • Used as a poly(ADP-ribose) polymerase inhibitor in cell-death studies.
  • Available in solid and solution formats for experimental flexibility.
  • High purity suitable for research applications.
  • Stable when stored under recommended temperature and light protection.
  • Molecular weight 136.15 g/mol and formula C7H8N2O.
Catalog No. 50-003-01766