Learn More
Medchemexpress LLC 4-Aminobenzo-12-crown-4 | 78554-68-4 | 98.0% | C12H17NO4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY78334
Encompass_Preferred
4-Aminobenzo-12-crown-4 is a chemical compound primarily utilized as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.
  • Used for the synthesis of various active compounds
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • For research use only
Catalog No. 50-003-58692