Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Aminobenzo-12-crown-4 | 78554-68-4 | 98.0% | C12H17NO4 | 5 G
SDP

Supplier:  Medchemexpress LLC HY783345G

Encompass_Preferred

4-Aminobenzo-12-crown-4 is a chemical compound that serves as a drug intermediate, utilized in the synthesis of various active compounds. It is identified for use in laboratory chemical applications and is supplied as a solid.

  • Functions as a drug intermediate
  • Used in the synthesis of various active compounds
  • Identified for laboratory chemical applications
  • Purity of 98.0%
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-34562


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.