Learn More
Medchemexpress LLC 4-Aminobenzohydrazide | 5351-17-7 | 99.2% | 100 MG

Supplier: Medchemexpress LLC HYB0880100MG
4-Aminobenzohydrazide (also known as p-Aminobenzohydrazide) is an irreversible myeloperoxidase (MPO) inhibitor with an IC50 of 0.3 μM. It induces an oxidative stress burst in neutrophils with an ROS IC50 of 43.6 μM and has been used in subacute stroke research. This compound has a molecular weight of 151.17 and the formula C7H9N3O. It appears as a light yellow to brown solid.
- Irreversible myeloperoxidase (MPO) inhibitor
- Induces oxidative stress burst in neutrophils
- Used in subacute stroke research
- Soluble in DMSO (≥ 25 mg/mL)
- Store at 4°C, protected from light
- Molecular formula: C7H9N3O
- Appearance: Light yellow to brown solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.