Learn More
Medchemexpress LLC 4-aminobenzoic acid | 150-13-0 | MFCD00007894 | 100.0% | 137.14 | C7H7NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1008R10MG
Encompass_Preferred
4-Aminobenzoic acid (PABA) analytical standard supplied in small pack sizes for research and analytical applications. It is used as an intermediate in folate biosynthesis and is provided with certificate-of-analysis-verified purity and molecular data for use as a calibration or reference standard.
  • Analytical standard intended for research and analytical applications.
  • Certificate of analysis (COA) confirms purity approximately 99.97%.
  • Molecular formula C7H7NO2, molecular weight 137.14.
  • Available in small pack sizes suitable for standards and calibration, including 10 mg.
  • Supplied as a solid suitable for analytical preparation and method validation.
Catalog No. 50-002-98209