Learn More
Selleck Chemical LLC 4-Aminobenzoic acid S4510-100mg
SDP
Supplier:  Selleck Chemical LLC S4510100MG
Encompass_Preferred
4-Aminobenzoic acid (para-Aminobenzoic acid) is an intermediate in the synthesis of tetrahydrofolic acid in many non-mammalian organisms including bacteria and fungi
Catalog No. 50-311-2186