Learn More
Apexbio Technology LLC 4-Aminobenzoic acid 150-13-0 5g
SDP
Supplier:  Apexbio Technology LLC B78695000
Encompass_Preferred
4-Aminobenzoic acid (CAS 150-13-0) is a small molecule functioning as a biosynthetic precursor in the formation of folate within bacteria plants and fungi It participates in microbial folate pathways by serving as a substrate for dihydropteroate synthase facilitating p-aminobenzoic acid incorporation into dihydropteroate a critical intermediate in folate synthesis Due to its involvement in this pathway 4-aminobenzoic acid is utilized in research investigating folate metabolism antimicrobial drug mechanisms and the biochemical processes underlying bacterial growth and resistance
Catalog No. 50-224-0585