Learn More
Medchemexpress LLC 4-aminobenzoic acid-d4 | 350820-01-8 | 99.5% | 141.16 | C7H3D4NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1008S10MG
Encompass_Preferred
4-Aminobenzoic acid-d4 is the deuterium-labeled isotopologue of 4-aminobenzoic acid supplied as a high-purity stable-isotope reagent for analytical and research use. It is used as a tracer and internal standard for quantitative analyses and supports common analytical techniques including NMR, GC-MS, and LC-MS.
  • High purity: 99.5%.
  • Deuterated internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable tracer for metabolic and folate pathway studies.
  • Available in small, precisely measured quantities for analytical workflows.
  • Chemical formula C7H3D4NO2; molecular weight 141.16.
Catalog No. 50-002-96708