Learn More
Medchemexpress LLC 4-Aminobenzoic acid potassium | 138-84-1 | 99.75% | 10 G
Supplier: Medchemexpress LLC HYB1008A10G
4-Aminobenzoic acid potassium is a modifier of catecholamine oxidation reactions. It is promising for research related to Parkinson's disease and hypertension. It is intended for research use only.
- Modifier of catecholamine oxidation reactions
- Promising for research related to Parkinson's disease and hypertension
- Solid appearance
- White to off-white color
- Soluble in H2O at 100 mg/mL