Learn More
Medchemexpress LLC 4-aminobenzonitrile-d4 | 1331866-32-0 | 122.16 | C7H2D4N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY41680S10MG
Encompass_Preferred
4-Aminobenzonitrile-d4 is the deuterium-labeled isotopologue of 4-aminobenzonitrile used as a stable isotope reagent and internal standard in analytical chemistry, tracer studies, and synthetic research. It is intended for laboratory-scale applications requiring isotopic differentiation and quantitative analysis.
  • Suitable as an internal standard for mass spectrometry.
  • Intended for use in analytical quantification and tracer studies.
  • Deuterium label facilitates discrimination from the native analyte.
  • Available in mg-scale quantities for laboratory workflows.
  • Molecular formula C7H2D4N2 and molecular weight 122.16 aid compound identification.
Catalog No. 50-002-99085