Learn More
Medchemexpress LLC 4-Aminobenzyl alcohol | 623-04-1 | 123.15 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007465
Encompass_Preferred
4-Aminobenzyl alcohol is a self-eliminating linker used to synthesize the fluorescent probe MB-Van1. This probe targets Vanin-1 and allows for real-time imaging of its activity in various inflammatory mouse models, including rheumatoid arthritis, drug-induced liver injury, and non-steroidal anti-inflammatory drug enteropathy. The probe undergoes an efficient self-elimination reaction after activation by the Vanin-1 enzyme, leading to significant signal amplification.
  • Can be used to synthesize fluorescent probes
  • Targets Vanin-1
  • Allows real-time imaging of Vanin-1's activity
  • Suitable for various inflammatory mouse models
  • Undergoes efficient self-elimination
  • Provides significant signal amplification
Catalog No. 50-004-49838