Learn More
Medchemexpress LLC 4-(aminomethyl)-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulfonamide | 2098346-67-7 | MFCD32198081 | 98.9% | 340.41 | C17H16N4O2S | 25 MG

Supplier: Medchemexpress LLC 25MGHYW037495
DCAF15 ligand-1 is a small-molecule research ligand for the E3 ligase DCAF15. It is commonly used as a building block in degrader and PROTAC synthesis and for studies of ubiquitin-proteasome pathway modulation. Analytical documentation (certificate of analysis) accompanies supplied material and provides recommended storage and handling instructions.
- Ligand for DCAF15 E3 ligase used in degrader synthesis and research.
- High purity as reported in certificate of analysis (98.87%).
- Molecular formula C17H16N4O2S and molecular weight 340.41 g/mol.
- CAS registry number 2098346-67-7 for unambiguous identification.
- Supplied as a small solid quantity suitable for synthesis (example package: 25 mg).
- Stability and storage recommendations provided in the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.