Learn More
Medchemexpress LLC 4-(Aminomethyl)pyridine | 3731-53-1 | 108.14 | 25 G

Supplier: Medchemexpress LLC 25GHYW007632
4-(Aminomethyl)pyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only and not sold to patients.
- Used as a biological material or organic compound for life science related research
- Appearance: colorless to yellow liquid
- Molecular formula: C6H8N2
- Molecular weight: 108.14
- Purity: 98.59%
- Store at -20°C, protect from light
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.