Learn More
Medchemexpress LLC 4-Aminonicotinic acid | 7418-65-7 | 138.12 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00255510G
Encompass_Preferred
4-Aminonicotinic acid is a versatile building block used for the synthesis of various pharmaceutical agents. It can also function as a substrate or inhibitor in research related to NAD(P)H metabolism and the activity of associated enzymes.
  • Can be used for the synthesis of pharmaceutical agents
  • Utilized as a substrate or inhibitor in research
  • Supports studies on NAD(P)H metabolism
  • Supports studies on related enzyme activity
Catalog No. 50-004-49502