Learn More
Medchemexpress LLC 4-Aminonicotinic acid | 7418-65-7 | 138.12 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0025555G
Encompass_Preferred
4-Aminonicotinic acid is a building block that can be used for the synthesis of pharmaceutical agents. It can also be utilized as a substrate or inhibitor in the research of NAD(P)H metabolism and the activity of related enzymes.
  • Used for the synthesis of pharmaceutical agents
  • Utilized as a substrate or inhibitor in NAD(P)H metabolism research
  • Utilized for activity of related enzymes
Catalog No. 50-004-49080