Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-arm-PEG-AC (MW 10000) | 95.0% | 10000 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY174968B

Encompass_Preferred

4-Arm-PEG-AC (MW 10000) is a polymer composed of a four-arm PEG core structure with acrylate groups attached to the ends of each PEG chain through a chemical reaction. This product is suitable for drug delivery applications.

  • Purity 95.0%
  • Molecular weight 10000 (average)
  • Solid, white to off-white appearance
  • Store powder at -20°C for 3 years
  • For research use only

Catalog No. 50-003-84118


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.