Learn More
Creative Pegworks 4-Arm PEG-Amine, MW 20k, 1 g
SDP
Supplier:  Creative Pegworks PSB4351G
Encompass
4-Arm PEG-Amine is a multiarm PEG derivative with amine groups at each terminal of the four arms connected to one pentaerythritol core. The reactive primary amine or NH2 group can rapidly react with activated carboxyl acid such as NHS ester to form stable amide bonds.
Catalog No. NC1850644