Learn More
Medchemexpress LLC 4-Arm PEG-Azide MW 50mg
SDP
Supplier:  Medchemexpress LLC HYW1048861D50MG
Encompass_Preferred
4-Arm PEG-Azide (MW 40000) (4-Arm PEG-N3 (MW 40000)) is a multi-arm PEG derivative with an azide group at each end of four arms attached to a pentaerythritol core[1]
Catalog No. 50-002-06874