Learn More
Medchemexpress LLC 4-arm-PEG-OH | 98.0% | 20000 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY174967C
Encompass_Preferred
4-Arm-PEG-OH (MW 20000) is a PEG derivative characterized by a four-arm structure and a hydroxyl (OH) functional group. This compound can react with functional groups present on biomolecules, such as proteins, peptides, and antibodies, through its hydroxyl groups. It is utilized in drug delivery applications.
  • Four-arm structure
  • Hydroxyl (OH) functional group
  • Reacts with functional groups on biomolecules
  • Suitable for drug delivery applications
Catalog No. 50-003-84392