Learn More
Medchemexpress LLC 4-Arm PEG-SCM MW 10 500mg
SDP
Supplier:  Medchemexpress LLC HYW1048855B500MG
Encompass_Preferred
4-Arm PEG-SCM (MW 10000) is a multi-arm PEG derivative that can be used for peptide modification and drug delivery[1]
Catalog No. 50-002-06873