Learn More
Cayman Chemical 4-AzIdo-L-phenylalnIn 1g
SDP
Supplier:  Cayman Chemical 416971 G
Encompass_Preferred
A derivative of the essential amino acid L-phenylalanine and a click chemistry reagent; has been used as a photocrosslinking agent and in spin labeling for the assessment of protein-protein interactions in vitro and in vivo
Catalog No. 50-294-4249