Learn More
Medchemexpress LLC 4-Azidobenzyl alcohol | 31499-54-4 | C7H7N3O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151840100MG
Encompass_Preferred
4-Azidobenzyl alcohol is a click chemistry reagent that contains an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. It is also capable of strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is for research use only.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Suitable for reactions with alkyne, DBCO, or BCN groups
Catalog No. 50-003-72803