Learn More
Medchemexpress LLC 4-Azidobenzyl alcohol | 31499-54-4 | C7H7N3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15184050MG
Encompass_Preferred
4-Azidobenzyl alcohol is a click chemistry reagent that contains an azide group. It is capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups.
  • Contains an azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN-containing molecules.
  • For research use only.
Catalog No. 50-003-75010